Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LNX1
Entrez Gene ID
84708
Gene Name
ligand of numb-protein X 1
Gene Aliases
LNX, MPDZ, PDZRN2
Chromosome Location
Chr.4: 53458264 - 53652524 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126328.2 NP_001119800.1 10 - 11 2335 101
NM_032622.2 NP_116011.2 9 - 10 2066 101
XM_005265785.4 XP_005265842.1 10 - 11 2493 101
XM_017008776.1 XP_016864265.1 11 - 12 2554 101
GenBank mRNA AF237782.1 - 9 - 10 1997 101
AK056823.1 - 10 - 11 2308 101
AK294611.1 - 10 - 11 1864 101
AY358326.1 - 9 - 10 1977 101
BC022983.1 - 10 - 11 2211 101
BC034737.1 - 9 - 10 1986 101
BC094772.1 - 6 - 7 1514 101

View Full Product Details