The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126328.2 | NP_001119800.1 | 3 - 4 | 906 | 120 |
NM_032622.2 | NP_116011.2 | 2 - 3 | 637 | 120 | |
XM_005265785.4 | XP_005265842.1 | 3 - 4 | 1064 | 120 | |
XM_017008776.1 | XP_016864265.1 | 4 - 5 | 1125 | 120 | |
GenBank mRNA | AF237782.1 | - | 2 - 3 | 568 | 120 |
AK056823.1 | - | 3 - 4 | 879 | 120 | |
AK294611.1 | - | 3 - 4 | 435 | 120 | |
AY358326.1 | - | 2 - 3 | 548 | 120 | |
BC022983.1 | - | 3 - 4 | 782 | 120 | |
BC034737.1 | - | 2 - 3 | 557 | 120 |