The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167604.1 | NP_001161076.1 | 11 - 12 | 1245 | 82 |
NM_001324128.1 | NP_001311057.1 | 9 - 10 | 1092 | 82 | |
NM_001324132.1 | NP_001311061.1 | 10 - 11 | 1156 | 82 | |
NM_001324133.1 | NP_001311062.1 | 11 - 12 | 1245 | 82 | |
NM_001324134.1 | NP_001311063.1 | 9 - 10 | 1076 | 82 | |
NM_001324136.1 | NP_001311065.1 | 11 - 12 | 1460 | 82 | |
NM_020383.3 | NP_065116.3 | 11 - 12 | 1245 | 82 | |
XM_011540134.1 | XP_011538436.1 | 11 - 12 | 1239 | 82 | |
XM_017016613.1 | XP_016872102.1 | 10 - 11 | 1095 | 82 | |
XM_017016614.1 | XP_016872103.1 | 12 - 13 | 1228 | 82 | |
XM_017016615.1 | XP_016872104.1 | 10 - 11 | 1112 | 82 | |
GenBank mRNA | AF272981.1 | - | 9 - 10 | 996 | 82 |
AK093693.1 | - | 9 - 10 | 1050 | 82 | |
AK223513.1 | - | 10 - 11 | 1056 | 82 | |
AK289436.1 | - | 11 - 12 | 1148 | 82 | |
AK304367.1 | - | 11 - 12 | 1138 | 82 | |
AL833411.1 | - | 11 - 12 | 1151 | 82 | |
BC005126.2 | - | 10 - 11 | 1034 | 82 | |
BC007579.1 | - | 11 - 12 | 1136 | 82 | |
BC013417.2 | - | 7 - 8 | 746 | 82 | |
BX537682.1 | - | 8 - 9 | 901 | 82 | |
CR456922.1 | - | 9 - 10 | 996 | 82 |