Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
XPNPEP1
Entrez Gene ID
7511
Gene Name
X-prolyl aminopeptidase 1
Gene Aliases
APP1, SAMP, XPNPEP, XPNPEPL, XPNPEPL1
Chromosome Location
Chr.10: 109864766 - 109923553 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167604.1 NP_001161076.1 19 - 20 1926 110
NM_001324128.1 NP_001311057.1 18 - 19 1845 110
NM_001324131.1 NP_001311060.1 20 - 21 2042 110
NM_001324132.1 NP_001311061.1 19 - 20 1909 110
NM_001324134.1 NP_001311063.1 18 - 19 1829 110
NM_001324135.1 NP_001311064.1 19 - 20 1947 110
NM_001324136.1 NP_001311065.1 20 - 21 2213 110
NM_020383.3 NP_065116.3 20 - 21 1998 110
XM_011540134.1 XP_011538436.1 19 - 20 1818 110
XM_017016613.1 XP_016872102.1 19 - 20 1848 110
XM_017016614.1 XP_016872103.1 21 - 22 1981 110
XM_017016615.1 XP_016872104.1 19 - 20 1865 110
GenBank mRNA AF195530.1 - 19 - 20 1825 110
AF272981.1 - 18 - 19 1749 110
AK093693.1 - 18 - 19 1803 110
AK095447.1 - 5 - 6 2044 110
AK223513.1 - 19 - 20 1809 110
AK289436.1 - 20 - 21 1901 110
AL833411.1 - 19 - 20 1832 110
BC005126.2 - 19 - 20 1787 110
BC007579.1 - 20 - 21 1889 110
BC013417.2 - 16 - 17 1499 110
BX537682.1 - 16 - 17 3843 110
CR456922.1 - 18 - 19 1749 110
X95762.1 - 18 - 19 1749 110

View Full Product Details