The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167604.1 | NP_001161076.1 | 8 - 9 | 865 | 133 |
NM_001324128.1 | NP_001311057.1 | 6 - 7 | 712 | 133 | |
NM_001324131.1 | NP_001311060.1 | 9 - 10 | 960 | 133 | |
NM_001324132.1 | NP_001311061.1 | 7 - 8 | 776 | 133 | |
NM_001324133.1 | NP_001311062.1 | 8 - 9 | 865 | 133 | |
NM_001324134.1 | NP_001311063.1 | 6 - 7 | 696 | 133 | |
NM_001324135.1 | NP_001311064.1 | 8 - 9 | 865 | 133 | |
NM_001324136.1 | NP_001311065.1 | 8 - 9 | 1080 | 133 | |
NM_020383.3 | NP_065116.3 | 8 - 9 | 865 | 133 | |
XM_011540134.1 | XP_011538436.1 | 8 - 9 | 859 | 133 | |
XM_017016613.1 | XP_016872102.1 | 7 - 8 | 715 | 133 | |
XM_017016614.1 | XP_016872103.1 | 9 - 10 | 848 | 133 | |
XM_017016615.1 | XP_016872104.1 | 7 - 8 | 732 | 133 | |
GenBank mRNA | AF195530.1 | - | 7 - 8 | 692 | 133 |
AF272981.1 | - | 6 - 7 | 616 | 133 | |
AK093693.1 | - | 6 - 7 | 670 | 133 | |
AK223513.1 | - | 7 - 8 | 676 | 133 | |
AK289436.1 | - | 8 - 9 | 768 | 133 | |
AK304367.1 | - | 8 - 9 | 758 | 133 | |
AL833411.1 | - | 8 - 9 | 771 | 133 | |
BC005126.2 | - | 7 - 8 | 654 | 133 | |
BC007579.1 | - | 8 - 9 | 756 | 133 | |
BC013417.2 | - | 4 - 5 | 366 | 133 | |
BX537682.1 | - | 5 - 6 | 521 | 133 | |
CR456922.1 | - | 6 - 7 | 616 | 133 | |
X95762.1 | - | 6 - 7 | 616 | 133 |