Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PYGL
Entrez Gene ID
5836
Gene Name
phosphorylase, glycogen, liver
Gene Aliases
GSD6
Chromosome Location
Chr.14: 50905217 - 50944530 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002863.4 NP_002854.3 2 - 3 473 88
GenBank mRNA AF046785.1 - 2 - 3 346 88
AF066858.1 - 2 - 3 346 88
AK225904.1 - 2 - 3 426 88
AK309333.1 - 2 - 3 399 88
AK313206.1 - 2 - 3 387 88
BC009895.2 - 2 - 3 416 88
BC065011.1 - 2 - 3 413 88
BC082229.1 - 2 - 3 417 88
BC095850.1 - 2 - 3 433 88
BC110791.1 - 2 - 3 415 88
DA287767.1 - 2 - 3 474 88
M14636.1 - 2 - 3 459 88
Y15233.1 - 2 - 3 397 88

View Full Product Details