Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EPB41L3
Entrez Gene ID
23136
Gene Name
erythrocyte membrane protein band 4.1 like 3
Gene Aliases
4.1B, DAL-1, DAL1
Chromosome Location
Chr.18: 5392381 - 5630666 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281534.1 NP_001268463.1 19 - 20 3011 90
NM_001281535.1 NP_001268464.1 21 - 22 2812 90
NM_012307.3 NP_036439.2 22 - 23 3677 90
XM_011525623.2 XP_011523925.1 22 - 23 3245 90
XM_011525626.2 XP_011523928.1 20 - 21 2811 90
XM_011525628.2 XP_011523930.1 21 - 22 3170 90
XM_011525632.2 XP_011523934.1 20 - 21 2724 90
XM_011525634.2 XP_011523936.1 19 - 20 2661 90
XM_011525637.2 XP_011523939.1 17 - 18 2559 90
XM_017025618.1 XP_016881107.1 19 - 20 2730 90
XM_017025620.1 XP_016881109.1 19 - 20 2688 90
XM_017025621.1 XP_016881110.1 18 - 19 2682 90
XM_017025622.1 XP_016881111.1 22 - 23 3191 90
XM_017025623.1 XP_016881112.1 18 - 19 2655 90
XM_017025624.1 XP_016881113.1 19 - 20 2643 90
XM_017025625.1 XP_016881114.1 21 - 22 2938 90
XM_017025627.1 XP_016881116.1 21 - 22 3164 90
XM_017025628.1 XP_016881117.1 20 - 21 3134 90
XM_017025629.1 XP_016881118.1 21 - 22 3116 90
XM_017025630.1 XP_016881119.1 21 - 22 2884 90
XM_017025632.1 XP_016881121.1 20 - 21 3089 90
XM_017025633.1 XP_016881122.1 20 - 21 2857 90
XM_017025635.1 XP_016881124.1 17 - 18 2544 90
XM_017025637.1 XP_016881126.1 20 - 21 3047 90
XM_017025639.1 XP_016881128.1 20 - 21 3035 90
XM_017025640.1 XP_016881129.1 19 - 20 3023 90
XM_017025641.1 XP_016881130.1 16 - 17 2478 90
XM_017025643.1 XP_016881132.1 19 - 20 2966 90
XM_017025644.1 XP_016881133.1 19 - 20 2957 90
XM_017025645.1 XP_016881134.1 22 - 23 3104 90
XM_017025646.1 XP_016881135.1 18 - 19 2930 90
XM_017025647.1 XP_016881136.1 21 - 22 3068 90
XM_017025648.1 XP_016881137.1 21 - 22 3029 90
XM_017025649.1 XP_016881138.1 22 - 23 3050 90
XM_017025650.1 XP_016881139.1 21 - 22 3023 90
XM_017025651.1 XP_016881140.1 20 - 21 2993 90
XM_017025652.1 XP_016881141.1 21 - 22 2975 90
XM_017025653.1 XP_016881142.1 21 - 22 2969 90
XM_017025654.1 XP_016881143.1 20 - 21 2948 90
XM_017025655.1 XP_016881144.1 20 - 21 2939 90
XM_017025657.1 XP_016881146.1 20 - 21 2906 90
XM_017025658.1 XP_016881147.1 20 - 21 2894 90
XM_017025659.1 XP_016881148.1 19 - 20 2870 90
XM_017025660.1 XP_016881149.1 20 - 21 2852 90
GenBank mRNA AB023204.1 - 22 - 23 3354 90
AF069072.1 - 19 - 20 2664 90
AK094952.1 - 9 - 10 1157 90
AK128531.1 - 21 - 22 2679 90
AK292583.1 - 22 - 23 2812 90
AK294690.1 - 10 - 11 1334 90
AK295453.1 - 11 - 12 1355 90
AK303679.1 - 11 - 12 1452 90
BC006141.1 - 20 - 21 2982 90
BC008377.1 - 8 - 9 1033 90

View Full Product Details