Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EPB41L3
Entrez Gene ID
23136
Gene Name
erythrocyte membrane protein band 4.1 like 3
Gene Aliases
4.1B, DAL-1, DAL1
Chromosome Location
Chr.18: 5392381 - 5630666 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281533.1 NP_001268462.1 3 - 4 612 116
NM_001281534.1 NP_001268463.1 3 - 4 793 116
NM_001281535.1 NP_001268464.1 5 - 6 594 116
NM_012307.3 NP_036439.2 3 - 4 793 116
XM_011525623.2 XP_011523925.1 3 - 4 793 116
XM_011525626.2 XP_011523928.1 2 - 3 395 116
XM_011525628.2 XP_011523930.1 3 - 4 793 116
XM_011525632.2 XP_011523934.1 2 - 3 395 116
XM_011525634.2 XP_011523936.1 2 - 3 395 116
XM_011525637.2 XP_011523939.1 2 - 3 395 116
XM_017025618.1 XP_016881107.1 2 - 3 395 116
XM_017025619.1 XP_016881108.1 2 - 3 395 116
XM_017025620.1 XP_016881109.1 2 - 3 395 116
XM_017025621.1 XP_016881110.1 2 - 3 395 116
XM_017025622.1 XP_016881111.1 3 - 4 793 116
XM_017025623.1 XP_016881112.1 2 - 3 395 116
XM_017025624.1 XP_016881113.1 2 - 3 395 116
XM_017025625.1 XP_016881114.1 3 - 4 561 116
XM_017025626.1 XP_016881115.1 2 - 3 395 116
XM_017025627.1 XP_016881116.1 3 - 4 793 116
XM_017025628.1 XP_016881117.1 3 - 4 793 116
XM_017025629.1 XP_016881118.1 3 - 4 793 116
XM_017025630.1 XP_016881119.1 3 - 4 561 116
XM_017025631.1 XP_016881120.1 2 - 3 395 116
XM_017025632.1 XP_016881121.1 3 - 4 793 116
XM_017025633.1 XP_016881122.1 3 - 4 561 116
XM_017025634.1 XP_016881123.1 2 - 3 395 116
XM_017025635.1 XP_016881124.1 2 - 3 395 116
XM_017025636.1 XP_016881125.1 3 - 4 793 116
XM_017025637.1 XP_016881126.1 3 - 4 793 116
XM_017025638.1 XP_016881127.1 2 - 3 395 116
XM_017025639.1 XP_016881128.1 3 - 4 793 116
XM_017025640.1 XP_016881129.1 3 - 4 793 116
XM_017025641.1 XP_016881130.1 2 - 3 395 116
XM_017025642.1 XP_016881131.1 3 - 4 793 116
XM_017025643.1 XP_016881132.1 3 - 4 793 116
XM_017025644.1 XP_016881133.1 3 - 4 793 116
XM_017025645.1 XP_016881134.1 3 - 4 652 116
XM_017025646.1 XP_016881135.1 3 - 4 793 116
XM_017025647.1 XP_016881136.1 3 - 4 652 116
XM_017025648.1 XP_016881137.1 3 - 4 652 116
XM_017025649.1 XP_016881138.1 3 - 4 652 116
XM_017025650.1 XP_016881139.1 3 - 4 652 116
XM_017025651.1 XP_016881140.1 3 - 4 652 116
XM_017025652.1 XP_016881141.1 3 - 4 652 116
XM_017025653.1 XP_016881142.1 3 - 4 652 116
XM_017025654.1 XP_016881143.1 3 - 4 652 116
XM_017025655.1 XP_016881144.1 3 - 4 652 116
XM_017025656.1 XP_016881145.1 3 - 4 652 116
XM_017025657.1 XP_016881146.1 3 - 4 652 116
XM_017025658.1 XP_016881147.1 3 - 4 652 116
XM_017025659.1 XP_016881148.1 3 - 4 652 116
XM_017025660.1 XP_016881149.1 3 - 4 652 116
GenBank mRNA AB023204.1 - 3 - 4 470 116
AF069072.1 - 3 - 4 450 116
AK128531.1 - 4 - 5 461 116
AK292583.1 - 5 - 6 594 116
AK297406.1 - 3 - 4 612 116
AK307904.1 - 3 - 4 714 116
AK314438.1 - 1 - 2 120 116
BC006141.1 - 3 - 4 764 116
BC008377.1 - 3 - 4 629 116

View Full Product Details