Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ICAM5
Entrez Gene ID
7087
Gene Name
intercellular adhesion molecule 5
Gene Aliases
TLCN, TLN
Chromosome Location
Chr.19: 10289352 - 10296778 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003259.3 NP_003250.3 3 - 4 737 112
XM_011528229.1 XP_011526531.1 4 - 5 809 112
GenBank mRNA AX746344.1 - 3 - 4 711 112
BC026338.1 - 3 - 4 735 112
BC030132.2 - 1 - 2 336 112
U72671.1 - 3 - 4 737 112

View Full Product Details