The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015387.4 | NP_056202.2 | 1 - 2 | 105 | 72 |
GenBank mRNA | AB015441.1 | - | 1 - 2 | 64 | 72 |
AF093825.1 | - | 1 - 2 | 80 | 72 | |
AF151853.1 | - | 1 - 2 | 80 | 72 | |
AJ580638.1 | - | 1 - 2 | 61 | 72 | |
AK027043.1 | - | 1 - 2 | 102 | 72 | |
AK292938.1 | - | 1 - 2 | 101 | 72 | |
AK307497.1 | - | 1 - 2 | 74 | 72 | |
AL080070.1 | - | 1 - 2 | 74 | 72 | |
BC005237.1 | - | 1 - 2 | 105 | 72 | |
CR457371.1 | - | 1 - 2 | 61 | 72 |