The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052955.2 | NP_443187.1 | 11 - 12 | 1979 | 71 |
XM_017021903.1 | XP_016877392.1 | 11 - 12 | 1977 | 71 | |
GenBank mRNA | AF363393.1 | - | 11 - 12 | 1979 | 71 |
BX112796.1 | - | 1 - 2 | 35 | 71 |