The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172779.1 | NP_001166250.1 | 5 - 6 | 760 | 165 |
NM_001172780.1 | NP_001166251.1 | 5 - 6 | 760 | 165 | |
NM_153353.4 | NP_699184.2 | 5 - 6 | 760 | 165 | |
XM_006713508.3 | XP_006713571.1 | 5 - 6 | 490 | 165 | |
GenBank mRNA | AK095125.1 | - | 5 - 6 | 505 | 165 |
AK293661.1 | - | 5 - 6 | 638 | 165 | |
AK295065.1 | - | 5 - 6 | 499 | 165 | |
AK302466.1 | - | 5 - 6 | 522 | 165 | |
BC032841.2 | - | 5 - 6 | 499 | 165 |