The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019010.2 | NP_061883.1 | 2 - 3 | 536 | 70 |
GenBank mRNA | AK312744.1 | - | 2 - 3 | 535 | 70 |
BC031559.1 | - | 2 - 3 | 520 | 70 |