Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRRC34
Entrez Gene ID
151827
Gene Name
leucine rich repeat containing 34
Gene Aliases
-
Chromosome Location
Chr.3: 169793006 - 169812904 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017005746.1 XP_016861235.1 1 - 2 334 161
GenBank mRNA AK095125.1 - 1 - 2 121 161

View Full Product Details