The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005704.4 | NP_005695.3 | 14 - 15 | 2451 | 68 |
XM_016999992.1 | XP_016855481.1 | 14 - 15 | 2426 | 68 | |
XM_016999993.1 | XP_016855482.1 | 14 - 15 | 2426 | 68 | |
XM_016999994.1 | XP_016855483.1 | 14 - 15 | 2426 | 68 | |
XM_016999995.1 | XP_016855484.1 | 9 - 10 | 1592 | 68 | |
GenBank mRNA | BM799833.1 | - | 1 - 2 | 99 | 68 |
X95712.1 | - | 15 - 16 | 2382 | 68 |