The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195001.1 | NP_001181930.1 | 25 - 26 | 3803 | 78 |
NM_005704.4 | NP_005695.3 | 26 - 27 | 3842 | 78 | |
NM_133177.3 | NP_573438.3 | 26 - 27 | 3830 | 78 | |
NM_133178.3 | NP_573439.2 | 25 - 26 | 3812 | 78 | |
XM_006710269.2 | XP_006710332.1 | 26 - 27 | 3805 | 78 | |
XM_016999992.1 | XP_016855481.1 | 27 - 28 | 3835 | 78 | |
XM_016999993.1 | XP_016855482.1 | 27 - 28 | 3826 | 78 | |
XM_016999994.1 | XP_016855483.1 | 26 - 27 | 3808 | 78 | |
XM_016999995.1 | XP_016855484.1 | 22 - 23 | 3001 | 78 | |
GenBank mRNA | AB208855.1 | - | 21 - 22 | 3106 | 78 |
BC146655.1 | - | 25 - 26 | 3770 | 78 | |
U71075.1 | - | 26 - 27 | 3704 | 78 | |
U73727.1 | - | 25 - 26 | 3803 | 78 | |
X95712.1 | - | 27 - 28 | 3764 | 78 |