The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278642.1 | NP_001265571.1 | 5 - 6 | 1111 | 81 |
NM_005711.4 | NP_005702.3 | 6 - 7 | 1141 | 81 | |
GenBank mRNA | AK312858.1 | - | 6 - 7 | 658 | 81 |
BC030828.1 | - | 6 - 7 | 1067 | 81 | |
BC053656.1 | - | 5 - 6 | 692 | 81 | |
BX648583.1 | - | 5 - 6 | 630 | 81 | |
U70312.1 | - | 6 - 7 | 758 | 81 |