The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077203.2 | NP_001070671.1 | 20 - 21 | 2707 | 79 |
NM_001282801.1 | NP_001269730.1 | 20 - 21 | 2704 | 79 | |
NM_001282802.1 | NP_001269731.1 | 20 - 21 | 2852 | 79 | |
NM_001282803.1 | NP_001269732.1 | 18 - 19 | 2459 | 79 | |
NM_020654.4 | NP_065705.3 | 21 - 22 | 2902 | 79 | |
XM_005247662.4 | XP_005247719.1 | 19 - 20 | 2495 | 79 | |
XM_011513038.2 | XP_011511340.1 | 21 - 22 | 2891 | 79 | |
XM_011513039.2 | XP_011511341.1 | 20 - 21 | 2816 | 79 | |
XM_011513040.2 | XP_011511342.1 | 20 - 21 | 2696 | 79 | |
XM_011513041.2 | XP_011511343.1 | 20 - 21 | 2693 | 79 | |
XM_011513042.2 | XP_011511344.1 | 19 - 20 | 2498 | 79 | |
XM_017006924.1 | XP_016862413.1 | 21 - 22 | 2779 | 79 | |
XM_017006925.1 | XP_016862414.1 | 21 - 22 | 2867 | 79 | |
XM_017006926.1 | XP_016862415.1 | 19 - 20 | 2605 | 79 | |
XM_017006927.1 | XP_016862416.1 | 19 - 20 | 2602 | 79 | |
GenBank mRNA | AB051494.1 | - | 14 - 15 | 1909 | 79 |
AF199458.1 | - | 20 - 21 | 2850 | 79 | |
AF217504.1 | - | 4 - 5 | 718 | 79 | |
AL136599.1 | - | 20 - 21 | 2730 | 79 | |
AL833577.1 | - | 20 - 21 | 2673 | 79 | |
BC129988.1 | - | 21 - 22 | 2829 | 79 | |
BX537943.1 | - | 18 - 19 | 2403 | 79 |