The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018067.1 | NP_001018077.1 | 7 - 8 | 1313 | 65 |
NM_001018068.1 | NP_001018078.1 | 7 - 8 | 1295 | 65 | |
NM_001018069.1 | NP_001018079.1 | 7 - 8 | 1268 | 65 | |
NM_015640.3 | NP_056455.3 | 7 - 8 | 1250 | 65 | |
GenBank mRNA | AF151813.1 | - | 7 - 8 | 1227 | 65 |
AK074970.1 | - | 7 - 8 | 1262 | 65 | |
AK130606.1 | - | 7 - 8 | 1196 | 65 | |
AK222955.1 | - | 7 - 8 | 1196 | 65 | |
AK223138.1 | - | 7 - 8 | 1196 | 65 | |
AK312256.1 | - | 7 - 8 | 1212 | 65 | |
AL080119.1 | - | 7 - 8 | 1195 | 65 | |
AY032853.1 | - | 7 - 8 | 877 | 65 | |
BC002488.2 | - | 7 - 8 | 1222 | 65 | |
BC003049.1 | - | 7 - 8 | 1221 | 65 | |
BC008045.2 | - | 7 - 8 | 1244 | 65 | |
BC017449.1 | - | 7 - 8 | 1181 | 65 | |
BC019273.1 | - | 7 - 8 | 1250 | 65 | |
BC020555.1 | - | 7 - 8 | 1227 | 65 | |
BC026916.1 | - | 7 - 8 | 1245 | 65 | |
BX647139.1 | - | 7 - 8 | 1225 | 65 | |
CR457383.1 | - | 7 - 8 | 1110 | 65 |