The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145898.3 | NP_665905.2 | 1 - 2 | 147 | 124 |
GenBank mRNA | BC093970.1 | - | 1 - 2 | 141 | 124 |
U67128.1 | - | 1 - 2 | 91 | 124 | |
U85767.1 | - | 1 - 2 | 106 | 124 |