Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C11orf1
Entrez Gene ID
64776
Gene Name
chromosome 11 open reading frame 1
Gene Aliases
-
Chromosome Location
Chr.11: 111879190 - 111884083 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022761.2 NP_073598.1 2 - 3 610 119
XM_011542957.2 XP_011541259.1 3 - 4 775 119
GenBank mRNA AJ250229.1 - 2 - 3 308 119
AK027152.1 - 2 - 3 293 119
BC020628.1 - 2 - 3 329 119
CR457378.1 - 2 - 3 273 119

View Full Product Details