The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020169.3 | NP_064554.3 | 5 - 6 | 785 | 116 |
GenBank mRNA | AF087851.1 | - | 5 - 6 | 648 | 116 |
AF282594.1 | - | 5 - 6 | 654 | 116 | |
AF282626.1 | - | 5 - 6 | 721 | 116 | |
BC005346.1 | - | 5 - 6 | 785 | 116 | |
BC008438.1 | - | 5 - 6 | 767 | 116 |