The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138705.2 | NP_619650.2 | 4 - 5 | 849 | 60 |
XM_005244729.3 | XP_005244786.2 | 4 - 5 | 1991 | 60 | |
XM_011540841.2 | XP_011539143.1 | 3 - 4 | 1874 | 60 | |
GenBank mRNA | AF490905.2 | - | 4 - 5 | 849 | 60 |
AY566229.1 | - | 4 - 5 | 849 | 60 |