The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257231.1 | NP_001244160.1 | 1 - 2 | 257 | 73 |
NM_001257237.1 | NP_001244166.1 | 1 - 2 | 257 | 73 | |
NM_001257240.2 | NP_001244169.1 | 1 - 2 | 257 | 73 | |
NM_001257241.2 | NP_001244170.1 | 1 - 2 | 257 | 73 | |
NM_001324293.1 | NP_001311222.1 | 1 - 2 | 257 | 73 | |
XM_011531031.1 | XP_011529333.1 | 1 - 2 | 257 | 73 | |
GenBank mRNA | AK302890.1 | - | 1 - 2 | 160 | 73 |
BF239206.1 | - | 1 - 2 | 211 | 73 | |
DA830058.1 | - | 1 - 2 | 248 | 73 | |
DB292609.1 | - | 1 - 2 | 257 | 73 | |
DN992019.1 | - | 1 - 2 | 252 | 73 |