The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152772.2 | NP_689985.1 | 6 - 7 | 960 | 146 |
XM_005268767.4 | XP_005268824.1 | 7 - 8 | 1110 | 146 | |
XM_005268768.4 | XP_005268825.1 | 6 - 7 | 1116 | 146 | |
XM_011538129.2 | XP_011536431.1 | 7 - 8 | 1878 | 146 | |
XM_011538130.2 | XP_011536432.1 | 7 - 8 | 1255 | 146 | |
XM_011538131.2 | XP_011536433.1 | 7 - 8 | 1113 | 146 | |
XM_011538132.2 | XP_011536434.1 | 6 - 7 | 995 | 146 | |
XM_017019127.1 | XP_016874616.1 | 6 - 7 | 1789 | 146 | |
XM_017019128.1 | XP_016874617.1 | 8 - 9 | 2025 | 146 | |
XM_017019129.1 | XP_016874618.1 | 8 - 9 | 1513 | 146 | |
XM_017019130.1 | XP_016874619.1 | 7 - 8 | 1061 | 146 | |
XM_017019131.1 | XP_016874620.1 | 6 - 7 | 833 | 146 | |
XM_017019132.1 | XP_016874621.1 | 8 - 9 | 1252 | 146 | |
XM_017019133.1 | XP_016874622.1 | 5 - 6 | 780 | 146 | |
GenBank mRNA | AK314055.1 | - | 6 - 7 | 900 | 146 |
BC033617.1 | - | 6 - 7 | 948 | 146 |