The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286974.1 | NP_001273903.1 | 17 - 18 | 2206 | 89 |
NM_003798.3 | NP_003789.1 | 17 - 18 | 2206 | 89 | |
XM_005252291.3 | XP_005252348.1 | 17 - 18 | 2174 | 89 | |
XM_017015250.1 | XP_016870739.1 | 16 - 17 | 1926 | 89 | |
XM_017015251.1 | XP_016870740.1 | 16 - 17 | 1926 | 89 | |
GenBank mRNA | AB451283.1 | - | 18 - 19 | 2054 | 89 |
AB451415.1 | - | 18 - 19 | 2054 | 89 | |
AF006070.1 | - | 17 - 18 | 2096 | 89 | |
AF030233.1 | - | 17 - 18 | 2098 | 89 | |
AF080071.1 | - | 17 - 18 | 2093 | 89 | |
AF135021.1 | - | 17 - 18 | 2097 | 89 | |
AI148526.1 | - | 2 - 3 | 89 | ||
AK022834.1 | - | 17 - 18 | 2139 | 89 | |
AK022926.1 | - | 17 - 18 | 2111 | 89 | |
AK123916.1 | - | 2 - 3 | 2911 | 89 | |
AK223230.1 | - | 17 - 18 | 2111 | 89 | |
AK314677.1 | - | 17 - 18 | 2110 | 89 | |
AL050016.1 | - | 10 - 11 | 918 | 89 | |
BC117208.1 | - | 17 - 18 | 2075 | 89 | |
BC117234.1 | - | 17 - 18 | 2075 | 89 | |
BU677451.1 | - | 2 - 3 | 89 | ||
U97067.1 | - | 17 - 18 | 2097 | 89 |