The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000124.3 | NP_000115.1 | 13 - 14 | 2779 | 98 |
GenBank mRNA | AB209504.1 | - | 7 - 8 | 6989 | 98 |
AK130100.1 | - | 8 - 9 | 1140 | 98 | |
AK303022.1 | - | 7 - 8 | 1028 | 98 | |
BC127104.1 | - | 13 - 14 | 2663 | 98 | |
CR749388.1 | - | 3 - 4 | 435 | 98 | |
L04791.1 | - | 13 - 14 | 2682 | 98 |