The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144622.2 | NP_653223.2 | 1 - 2 | 353 | 77 |
GenBank mRNA | AK057496.1 | - | 1 - 2 | 353 | 77 |
BC111794.1 | - | 1 - 2 | 332 | 77 | |
DB025590.1 | - | 1 - 2 | 353 | 77 |