Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DCST2
Entrez Gene ID
127579
Gene Name
DC-STAMP domain containing 2
Gene Aliases
-
Chromosome Location
Chr.1: 155018513 - 155033781 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144622.2 NP_653223.2 4 - 5 822 65
XM_011509188.2 XP_011507490.1 2 - 3 285 65
GenBank mRNA AK057496.1 - 4 - 5 822 65
AK097468.1 - 2 - 3 236 65
BC111794.1 - 4 - 5 801 65

View Full Product Details