Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DCST2
Entrez Gene ID
127579
Gene Name
DC-STAMP domain containing 2
Gene Aliases
-
Chromosome Location
Chr.1: 155018513 - 155033781 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144622.2 NP_653223.2 9 - 10 1591 65
XM_011509188.2 XP_011507490.1 7 - 8 1054 65
XM_011509189.2 XP_011507491.1 4 - 5 1443 65
GenBank mRNA AK057496.1 - 9 - 10 1591 65
AK097468.1 - 7 - 8 1005 65
AK097993.1 - 4 - 5 1263 65
BC111794.1 - 9 - 10 1570 65

View Full Product Details