The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173591.3 | NP_775862.3 | 44 - 45 | 5386 | 98 |
XM_005268802.3 | XP_005268859.1 | 48 - 49 | 6187 | 98 | |
XM_011538191.1 | XP_011536493.1 | 45 - 46 | 5463 | 98 | |
XM_011538192.2 | XP_011536494.1 | 42 - 43 | 5404 | 98 | |
XM_011538193.1 | XP_011536495.1 | 42 - 43 | 5323 | 98 |