Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNGB1
Entrez Gene ID
1258
Gene Name
cyclic nucleotide gated channel beta 1
Gene Aliases
CNCG2, CNCG3L, CNCG4, CNG4, CNGB1B, GAR1, GARP, GARP2, RCNC2, RCNCb, RCNCbeta, RP45
Chromosome Location
Chr.16: 57882340 - 57971116 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135639.1 NP_001129111.1 1 - 2 59 124
NM_001286130.1 NP_001273059.1 1 - 2 59 124
NM_001297.4 NP_001288.3 1 - 2 59 124
GenBank mRNA BQ638084.1 - 1 - 2 55 124
U58837.1 - 1 - 2 51 124

View Full Product Details