The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135639.1 | NP_001129111.1 | 2 - 3 | 223 | 60 |
NM_001286130.1 | NP_001273059.1 | 2 - 3 | 223 | 60 | |
NM_001297.4 | NP_001288.3 | 2 - 3 | 223 | 60 | |
GenBank mRNA | AF042498.1 | - | 2 - 3 | 228 | 60 |
BQ638084.1 | - | 2 - 3 | 219 | 60 | |
U18945.1 | - | 2 - 3 | 226 | 60 | |
U58837.1 | - | 2 - 3 | 215 | 60 |