The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135639.1 | NP_001129111.1 | 6 - 7 | 476 | 103 |
NM_001286130.1 | NP_001273059.1 | 6 - 7 | 476 | 103 | |
NM_001297.4 | NP_001288.3 | 6 - 7 | 476 | 103 | |
GenBank mRNA | AF042498.1 | - | 6 - 7 | 481 | 103 |
BQ638084.1 | - | 6 - 7 | 472 | 103 | |
U18945.1 | - | 6 - 7 | 479 | 103 | |
U58837.1 | - | 6 - 7 | 468 | 103 |