The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135639.1 | NP_001129111.1 | 9 - 10 | 650 | 74 |
NM_001297.4 | NP_001288.3 | 9 - 10 | 650 | 74 | |
GenBank mRNA | U18945.1 | - | 9 - 10 | 653 | 74 |
U58837.1 | - | 9 - 10 | 642 | 74 |