Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC66
Entrez Gene ID
285331
Gene Name
coiled-coil domain containing 66
Gene Aliases
-
Chromosome Location
Chr.3: 56557125 - 56621837 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012506.4 NP_001012524.4 10 - 11 1469 89
NM_001141947.1 NP_001135419.1 10 - 11 1488 89
NR_024460.1 10 - 11 1469 89
XM_005265081.3 XP_005265138.1 10 - 11 1488 89
XM_005265082.3 XP_005265139.1 10 - 11 1467 89
XM_005265083.3 XP_005265140.1 10 - 11 1359 89
XM_005265084.3 XP_005265141.1 10 - 11 1482 89
XM_011533614.2 XP_011531916.1 9 - 10 1493 89
XM_011533615.2 XP_011531917.1 9 - 10 1503 89
XM_011533616.2 XP_011531918.1 10 - 11 1427 89
XM_011533617.2 XP_011531919.1 11 - 12 1585 89
XM_011533619.2 XP_011531921.1 11 - 12 1488 89
XM_017006229.1 XP_016861718.1 10 - 11 1467 89
XM_017006230.1 XP_016861719.1 9 - 10 1471 89
XM_017006231.1 XP_016861720.1 9 - 10 1492 89
XM_017006232.1 XP_016861721.1 10 - 11 1359 89
XM_017006233.1 XP_016861722.1 10 - 11 1338 89
XM_017006234.1 XP_016861723.1 10 - 11 1527 89
XM_017006235.1 XP_016861724.1 11 - 12 1585 89
XM_017006236.1 XP_016861725.1 12 - 13 1580 89
XM_017006237.1 XP_016861726.1 12 - 13 1617 89
XM_017006238.1 XP_016861727.1 13 - 14 1758 89
XM_017006239.1 XP_016861728.1 7 - 8 818 89
XM_017006240.1 XP_016861729.1 11 - 12 1539 89
XM_017006241.1 XP_016861730.1 12 - 13 1498 89
XM_017006242.1 XP_016861731.1 11 - 12 1361 89
XM_017006243.1 XP_016861732.1 10 - 11 1454 89
XM_017006244.1 XP_016861733.1 11 - 12 1590 89
XM_017006245.1 XP_016861734.1 9 - 10 1275 89
GenBank mRNA AL832692.1 - 10 - 11 1408 89
BC047509.1 - 10 - 11 1437 89
BC132827.1 - 10 - 11 1455 89

View Full Product Details