Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CCDC66
- Entrez Gene ID
-
285331
- Gene Name
- coiled-coil domain containing 66
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 56557125 - 56621837 on Build GRCh38
View Full Product Details