The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001162491.1 | NP_001155963.1 | 4 - 5 | 500 | 72 |
NR_103708.1 | 4 - 5 | 500 | 72 | ||
NR_103709.1 | 4 - 5 | 500 | 72 | ||
XM_011530950.2 | XP_011529252.1 | 4 - 5 | 1197 | 72 | |
GenBank mRNA | AK301813.1 | - | 4 - 5 | 500 | 72 |
BC140808.1 | - | 4 - 5 | 574 | 72 | |
BC144603.1 | - | 4 - 5 | 574 | 72 | |
CD300373.1 | - | 1 - 2 | 224 | 72 | |
DY655154.1 | - | 1 - 2 | 72 |