The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015164.2 | NP_055979.2 | 17 - 18 | 2845 | 94 |
XM_005245790.3 | XP_005245847.1 | 16 - 17 | 2980 | 94 | |
XM_005245791.4 | XP_005245848.1 | 12 - 13 | 2113 | 94 | |
XM_017000757.1 | XP_016856246.1 | 17 - 18 | 2752 | 94 | |
XM_017000758.1 | XP_016856247.1 | 16 - 17 | 2692 | 94 | |
GenBank mRNA | AB020649.1 | - | 17 - 18 | 2621 | 94 |
AL137297.1 | - | 1 - 2 | 149 | 94 | |
BC008002.1 | - | 9 - 10 | 882 | 94 | |
BC016488.1 | - | 9 - 10 | 1527 | 94 | |
BC030545.1 | - | 8 - 9 | 825 | 94 | |
BC040441.1 | - | 17 - 18 | 2848 | 94 | |
BC042103.1 | - | 9 - 10 | 1175 | 94 | |
BC068599.1 | - | 9 - 10 | 1200 | 94 |