The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000175.3 | NP_000166.2 | 1 - 2 | 263 | 66 |
NM_001184722.1 | NP_001171651.1 | 2 - 3 | 410 | 66 | |
NM_001289789.1 | NP_001276718.1 | 2 - 3 | 410 | 66 | |
NM_001289790.1 | NP_001276719.1 | 1 - 2 | 263 | 66 | |
XM_005258764.2 | XP_005258821.1 | 2 - 3 | 229 | 66 | |
XM_006723148.2 | XP_006723211.1 | 2 - 3 | 180 | 66 | |
XM_011526754.2 | XP_011525056.1 | 3 - 4 | 645 | 66 | |
GenBank mRNA | AK129884.1 | - | 2 - 3 | 194 | 66 |
AK293446.1 | - | 1 - 2 | 139 | 66 | |
AK294396.1 | - | 2 - 3 | 358 | 66 | |
AK301103.1 | - | 1 - 2 | 148 | 66 | |
BC004982.1 | - | 1 - 2 | 131 | 66 | |
DC299841.1 | - | 1 - 2 | 263 | 66 | |
DC342043.1 | - | 2 - 3 | 410 | 66 | |
K03515.1 | - | 1 - 2 | 138 | 66 | |
X16539.1 | - | 1 - 2 | 126 | 66 |