The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005816.4 | NP_005807.1 | 11 - 12 | 1557 | 120 |
NM_198196.2 | NP_937839.1 | 12 - 13 | 1605 | 120 | |
NR_134917.1 | 11 - 12 | 1554 | 117 | ||
XM_006713469.3 | XP_006713532.1 | 12 - 13 | 1563 | 117 | |
XM_006713470.3 | XP_006713533.1 | 11 - 12 | 1515 | 117 | |
XM_017005521.1 | XP_016861010.1 | 11 - 12 | 1377 | 120 | |
XM_017005522.1 | XP_016861011.1 | 9 - 10 | 1254 | 120 | |
GenBank mRNA | AK315237.1 | - | 11 - 12 | 1439 | 120 |
AL833681.1 | - | 12 - 13 | 1452 | 120 |