The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005816.4 | NP_005807.1 | 12 - 13 | 1642 | 79 |
NM_198196.2 | NP_937839.1 | 13 - 14 | 1690 | 79 | |
NR_134917.1 | 12 - 13 | 1639 | 79 | ||
XM_006713469.3 | XP_006713532.1 | 13 - 14 | 1648 | 79 | |
XM_006713470.3 | XP_006713533.1 | 12 - 13 | 1600 | 79 | |
XM_017005521.1 | XP_016861010.1 | 12 - 13 | 1462 | 79 | |
XM_017005522.1 | XP_016861011.1 | 10 - 11 | 1339 | 79 | |
GenBank mRNA | AK290719.1 | - | 12 - 13 | 1525 | 79 |
AK315237.1 | - | 12 - 13 | 1524 | 79 | |
AL833681.1 | - | 13 - 14 | 1537 | 79 |