The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005816.4 | NP_005807.1 | 13 - 14 | 1773 | 72 |
NM_198196.2 | NP_937839.1 | 14 - 15 | 1821 | 72 | |
NR_134917.1 | 13 - 14 | 1770 | 72 | ||
XM_006713469.3 | XP_006713532.1 | 14 - 15 | 1779 | 72 | |
XM_006713470.3 | XP_006713533.1 | 13 - 14 | 1731 | 72 | |
XM_017005521.1 | XP_016861010.1 | 13 - 14 | 1593 | 72 | |
XM_017005522.1 | XP_016861011.1 | 11 - 12 | 1470 | 72 | |
GenBank mRNA | AK290719.1 | - | 13 - 14 | 1656 | 72 |
AK315237.1 | - | 13 - 14 | 1655 | 72 | |
AL833681.1 | - | 14 - 15 | 1668 | 72 |