Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NCS1
Entrez Gene ID
23413
Gene Name
neuronal calcium sensor 1
Gene Aliases
FLUP, FREQ
Chromosome Location
Chr.9: 130172578 - 130237304 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014286.3 NP_055101.2 2 - 3 176 91
GenBank mRNA AF134479.1 - 2 - 3 157 91
AF186409.1 - 2 - 3 103 91
BC004856.2 - 2 - 3 176 91
X84048.1 - 2 - 3 104 91

View Full Product Details