Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NCS1
Entrez Gene ID
23413
Gene Name
neuronal calcium sensor 1
Gene Aliases
FLUP, FREQ
Chromosome Location
Chr.9: 130172578 - 130237304 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128826.1 NP_001122298.1 5 - 6 352 88
NM_014286.3 NP_055101.2 5 - 6 483 88
GenBank mRNA AF134479.1 - 5 - 6 464 88
AF186409.1 - 5 - 6 410 88
BC004856.2 - 5 - 6 483 88
BQ880305.1 - 5 - 6 352 88
DA092531.1 - 5 - 6 351 88
X84048.1 - 5 - 6 411 88

View Full Product Details