The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003855.3 | NP_003846.1 | 2 - 3 | 206 | 130 |
XM_005264039.3 | XP_005264096.1 | 1 - 2 | 88 | 130 | |
XM_005264040.3 | XP_005264097.1 | 1 - 2 | 88 | 130 | |
XM_011512099.1 | XP_011510401.1 | 1 - 2 | 88 | 130 | |
XM_017005181.1 | XP_016860670.1 | 1 - 2 | 88 | 130 | |
GenBank mRNA | DR006178.1 | - | 3 - 4 | 416 | 130 |