The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013393.1 | NP_037525.1 | 2 - 3 | 325 | 78 |
GenBank mRNA | AF093415.1 | - | 2 - 3 | 325 | 78 |
BC017106.2 | - | 1 - 2 | 288 | 78 | |
BC114514.1 | - | 2 - 3 | 310 | 78 | |
BC114564.1 | - | 2 - 3 | 279 | 78 | |
CR749618.1 | - | 2 - 3 | 334 | 78 | |
EU794658.1 | - | 2 - 3 | 325 | 78 |