Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC4A1
Entrez Gene ID
6521
Gene Name
solute carrier family 4 member 1 (Diego blood group)
Gene Aliases
AE1, BND3, CD233, CHC, DI, EMPB3, EPB3, FR, RTA1A, SAO, SPH4, SW, WD, WD1, WR
Chromosome Location
Chr.17: 44248390 - 44268161 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000342.3 NP_000333.1 18 - 19 2636 68
XM_005257593.4 XP_005257650.1 16 - 17 2404 68
XM_011525129.1 XP_011523431.1 17 - 18 2573 68
GenBank mRNA AB526447.1 - 16 - 17 2490 68
BC096106.3 - 17 - 18 2549 68
BC096107.3 - 17 - 18 2549 68
BC099628.3 - 17 - 18 2549 68
BC099629.3 - 17 - 18 2549 68
BC101570.1 - 18 - 19 2573 68
BC101574.1 - 17 - 18 2569 68
DQ419529.1 - 17 - 18 2555 68
GQ981383.1 - 17 - 18 2530 68
GQ981384.1 - 17 - 18 2530 68
GQ981385.1 - 12 - 13 1432 68
GQ981386.1 - 17 - 18 2530 68
GQ981387.1 - 17 - 18 2530 68
M27819.1 - 18 - 19 2601 68
X12609.1 - 18 - 19 2637 68

View Full Product Details