The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017957.2 | NP_060427.2 | 3 - 4 | 1114 | 145 |
GenBank mRNA | AF324241.1 | - | 2 - 3 | 679 | 145 |
AK000785.1 | - | 3 - 4 | 1063 | 145 | |
AK291658.1 | - | 3 - 4 | 1063 | 145 | |
AK292977.1 | - | 3 - 4 | 1063 | 145 | |
AK300642.1 | - | 4 - 5 | 562 | 145 | |
BC110605.2 | - | 2 - 3 | 809 | 145 |