Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EPN3
Entrez Gene ID
55040
Gene Name
epsin 3
Gene Aliases
-
Chromosome Location
Chr.17: 50532687 - 50543750 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017957.2 NP_060427.2 6 - 7 1413 73
GenBank mRNA AF324241.1 - 5 - 6 978 73
AK000785.1 - 6 - 7 1362 73
AK291658.1 - 6 - 7 1362 73
AK292977.1 - 6 - 7 1362 73
AK296346.1 - 6 - 7 1199 73
AK300642.1 - 7 - 8 861 73
BC110602.2 - 5 - 6 945 73
BC110605.2 - 5 - 6 1151 73

View Full Product Details